Learn More
Eppendorf™ Rotor Lids
Product Code. 10269753 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
A-2-MTP
Centrifuge 5427R, FA-45-24-11-Kit
Centrifuge 5427R, FA-45-30-11
F-34-6-38
F-45-17-17-Cyro
F-45-30-11
F-45-48-11
F-45-64-5-PCR
FA-45-16-17
FA-45-24-11
FA-45-24-11-HS
FA-45-24-11-Kit
FA-45-30-11
FA-45-48-11
S-24-11-AT
Material:
Aluminum
Aluminum, Coated
Polypropylene
Description:
Aerosol-tight
Aerosol-tight, QuickLock
Coated
Non-aerosol-tight
Wind shield upper shell
Unit Size:
Each
Product Code. For Use With (Equipment) Material Description unitSize
10269753 FA-45-24-11-Kit Aluminum Aerosol-tight Each
12334948 A-2-MTP Aluminum Wind shield upper shell Each
15234536 Centrifuge 5427R, FA-45-24-11-Kit Aluminum Aerosol-tight Each
10791755 F-34-6-38 Aluminum Non-aerosol-tight Each
15244536 Centrifuge 5427R, FA-45-30-11 Aluminum Aerosol-tight Each
10197523 F-45-17-17-Cyro Polypropylene Non-aerosol-tight Each
15254536 F-45-30-11 Polypropylene Non-aerosol-tight Each
10005693 F-45-30-11 Polypropylene Coated Each
10782135 F-45-64-5-PCR Aluminum Non-aerosol-tight Each
15224536 FA-45-24-11 Aluminum Aerosol-tight Each
13226589 FA-45-16-17 Aluminum Aerosol-tight Each
10279323 FA-45-24-11-HS Aluminum Aerosol-tight Each
13246938 F-45-48-11 Polypropylene Non-aerosol-tight Each
13286948 FA-45-24-11-Kit Aluminum Aerosol-tight, QuickLock Each
10072383 FA-45-30-11 Aluminum Aerosol-tight Each
10496132 FA-45-30-11 Aluminum Non-aerosol-tight Each
13216958 FA-45-30-11 Aluminum Aerosol-tight, QuickLock Each
10076213 FA-45-30-11 Aluminum, Coated Aerosol-tight Each
13246958 FA-45-48-11 Aluminum Aerosol-tight, QuickLock Each
15274536 FA-45-48-11 Aluminum Aerosol-tight Each
15284536 FA-45-48-11 Polypropylene Non-aerosol-tight Each
13236948 S-24-11-AT Aluminum Aerosol-tight Each
22 options
Product Code. 10269753
Brand: Eppendorf™ 5427704000
Designed for use with Eppendorf Rotors
  • Replacement rotor lids
  • Aerosol-tight and aluminum lids available
  • Available in a variety of sizes
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Lid
Material Aluminum
For Use With (Equipment) FA-45-24-11-Kit
Description Aerosol-tight