missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human Monocyte Enrichment Cocktail

Product Code. 17167661
Encompass
Change view
Click to view available options
Quantity:
200 mL whole blood
40 mL whole blood
Unit Size:
1 set
200mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17167661 40 mL whole blood 1 set
15939945 200 mL whole blood 200mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17167661 Supplier STEMCELL Technologies Supplier No. 15028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity isolation of untouched monocytes

  • The RosetteSep™ Human Monocyte Enrichment Cocktail is designed to isolate monocytes from whole blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes (TAC) recognizing non-monocyte cells and red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified monocytes are present as a highly enriched population at the interface between the plasma and the buoyant density medium

Specifications

Cell Type Monocytes
For Use With (Application) Cell Isolation
Quantity 40 mL whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.