missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human Monocyte Depletion Cocktail

Product Code. 17107711
Encompass
Change view
Click to view available options
Quantity:
200 mL whole blood
40 mL whole blood
Unit Size:
1 set
Product Code. Quantity unitSize
17107711 200 mL whole blood 1 set
17147701 40 mL whole blood 1 set
2 options
This item is not returnable. View return policy

Product Code. 17107711

Brand: STEMCELL Technologies 15668

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity depletion of monocytes

  • The RosetteSep™ Human Monocyte Depletion Cocktail is designed to deplete monocytes from whole blood
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD36 and glycophorin A on red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The monocyte depleted fraction is present as a highly enriched population at the interface between the plasma and the buoyant density medium

Specifications

Cell Type Monocytes
For Use With (Application) Cell Isolation
Quantity 200 mL whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.