Learn More
MP Biomedicals™ Rogers and Harper Salt Mixture
Shop All MP Biomedicals Products
missing translation for 'orderingAttributeHoverText'
Quantity:
2 kg
10 kg
50 kg
missing translation for 'unitSize'
10kg
2kg
50kg
Description
- Salt Mixture
- For research animal diet
- Powder
Specifications
Specifications
| Product Type | Animal Diet |
| For Use With (Application) | Research animal diet |
| Quantity | 50 kg |
| Storage Requirements | Room Temperature |