Learn More
MP Biomedicals™ Rogers and Harper Salt Mixture
Product Code. 11415652 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415652 2 kg 2kg
11425652 10 kg 10kg
11435652 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11415652 Supplier MP Biomedicals™ Supplier No. 0290284202
 Restricted Item
Estimated Shipment: 08-12-2026
Salt mixture for research animal diet.
  • Salt Mixture
  • For research animal diet
  • Powder
Specifications
Product Type Animal Diet
For Use With (Application) Research animal diet
Quantity 2 kg
Storage Requirements Room Temperature