missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RoboSep™ Human CB CD34 Positive Selection Kit II

Product Code. 17187811
Encompass
Change view
Click to view available options
Quantity:
1000 mL cord blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187811 1000 mL cord blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187811 Supplier STEMCELL Technologies Supplier No. 17896RF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips

  • Isolate highly purified CD34+ cells from fresh whole umbilical cord blood using a simple, two-step procedure
  • Hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Cord Blood CD34 Pre-Enrichment Cocktail (15896C), through the use of antibodies to remove cells expressing T cell, B cell, myeloid cell and platelet surface markers
  • CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18096C), which contains an antibody recognizing CD34
  • If isolating CD34+ cells from fresh blood or buffy coat the Complete Kit for Human Whole Blood CD34+ Cells (Catalog #15086) is recommended
  • If isolating CD34+ cells from other samples, including fresh or previously frozen mobilized peripheral blood or bone marrow mononuclear cells, or from previously frozen cord blood mononuclear cells, we recommend using the EasySep™ Human CD34 Positive Selection Kit (Catalog #17856)

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Isolation
Quantity 1000 mL cord blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.