Description
Use the Thermo Scientific™ Rheodyne™ Suction Needle Adapter for suction loading of samples into the Rheodyne 9725 and 9725i Sample Injectors for high-performance liquid chromatography (HPLC),
Specifications
Specifications
| Type | Suction Needle Adapter |
| For Use With (Application) | Liquid Chromatography |
| Description | Suction Needle Adapter |
| For Use With (Equipment) | Rheodyne Injector Models 9725 and 9725i |