Learn More
Thermo Scientific™ Rheodyne™ Stators
Product Code. 10179324 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector Models 7725
Rheodyne Injectors 7000/7010/7030/7040/7125
Rheodyne Injectors 7010-087/7125-081
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7410/7413
Rheodyne Injectors 8125/8126
Rheodyne Injectors 9010/9030/9125
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
10179324 Rheodyne Injectors 7060/7066 Each
10056264 Rheodyne Injectors 7000/7010/7030/7040/7125 Each
10775126 Rheodyne Injectors 7010-087/7125-081 Each
10698795 Rheodyne Injectors 7410/7413 Each
10795526 Rheodyne Injector Models 7725 Each
10713406 Rheodyne Injectors 8125/8126 Each
10353055 Rheodyne Injectors 9010/9030/9125 Each
7 options
Product Code. 10179324
Brand: Thermo Scientific™ 7060039
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use these Thermo Scientific; Rheodyne; Stators for your Rheodyne Sample Injectors.
Use Thermo Scientific™ Rheodyne™ Stators with popular Rheodyne injector models used in high-performance liquid chromatography (HPLC) systems.
TRUSTED_SUSTAINABILITY
Specifications
Type Stators
Description For Rheodyne 7060/7066
For Use With (Equipment) Rheodyne Injectors 7060/7066