Learn More
Thermo Scientific™ Rheodyne™ Sample Loops
Product Code. 10678715
Click to view available options
Volume (Metric):
2 μL
5 μL
10 μL
20 μL
50 μL
100 μL
200 μL
500 μL
1 mL
5 mL
For Use With (Equipment):
Rheodyne Injector Model 8125
Rheodyne Injectors 7010/7125
Rheodyne Injectors 7725/7725i
Rheodyne Injectors 9010/9725
Rheodyne Injectors 9725/9725i
Diameter (Metric) Inner:
0.18 mm
0.20 mm
0.25 mm
0.30 mm
0.51 mm
0.76 mm
0.8 mm, 0.5 mm, and 0.2 mm
1.0 mm
Unit Size:
Each
Product Code. 10678715
Brand: Thermo Scientific™ 8021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
TRUSTED_SUSTAINABILITY
Specifications
Type Sample Loop
Description For Rheodyne 8125; 10 uL; 0.20 mm ID
For Use With (Equipment) Rheodyne Injector Model 8125
Diameter (English) Inner 0.008 in.
Diameter (Metric) Inner 0.20 mm
Volume (Metric) 10 μL