Learn More
- Home
- Products
- Chromatography Systems
- Liquid Chromatography Instruments
- Individual HPLC Components and Accessories
- HPLC Valve and Injector Accessories
- Thermo Scientific™ Rheodyne™ Sample Loops
Thermo Scientific™ Rheodyne™ Sample Loops
Product Code. 10575625| Product Code. | Volume (Metric) | For Use With (Equipment) | Diameter (Metric) Inner | unitSize |
|---|---|---|---|---|
| 10575625 | 20 μL | Rheodyne Injector Model 8125 | 0.25 mm | Each |
| 10455163 | 5 μL | Rheodyne Injectors 7010/7125 | 0.18 mm | Each |
| 10621334 | 10 μL | Rheodyne Injectors 7010/7125 | 0.30 mm | Each |
| 10100563 | 100 μL | Rheodyne Injectors 7010/7125 | 0.51 mm | Each |
| 10776524 | 200 μL | Rheodyne Injectors 7010/7125 | 0.76 mm | Each |
| 10682864 | 500 μL | Rheodyne Injectors 7010/7125 | 0.76 mm | Each |
| 10465173 | 5 mL | Rheodyne Injectors 7010/7125 | 1.0 mm | Each |
| 10344473 | 2 μL | Rheodyne Injectors 9725/9725i | 0.8 mm, 0.5 mm, and 0.2 mm | Each |
| 10794794 | 5 μL | Rheodyne Injectors 7725/7725i | 0.18 mm | Each |
| 10766714 | 10 μL | Rheodyne Injectors 7725/7725i | 0.30 mm | Each |
| 10466113 | 20 μL | Rheodyne Injectors 7725/7725i | 0.30 mm | Each |
| 10620184 | 50 μL | Rheodyne Injectors 7725/7725i | 0.51 mm | Each |
| 10408104 | 5 μL | Rheodyne Injector Model 8125 | 0.20 mm | Each |
| 10678715 | 10 μL | Rheodyne Injector Model 8125 | 0.20 mm | Each |
| 10147854 | 50 μL | Rheodyne Injector Model 8125 | 0.30 mm | Each |
| 10269364 | 5 μL | Rheodyne Injectors 9725/9725i | 0.18 mm | Each |
| 10219324 | 10 μL | Rheodyne Injectors 9725/9725i | 0.25 mm | Each |
| 10555615 | 20 μL | Rheodyne Injectors 9725/9725i | 0.25 mm | Each |
| 10311255 | 50 μL | Rheodyne Injectors 9725/9725i | 0.51 mm | Each |
| 10229414 | 100 μL | Rheodyne Injectors 9010/9725 | 0.51 mm | Each |
| 10651396 | 200 μL | Rheodyne Injectors 9010/9725 | 0.51 mm | Each |
| 10774057 | 500 μL | Rheodyne Injectors 9010/9725 | 0.76 mm | Each |
| 10321255 | 1 mL | Rheodyne Injectors 9010/9725 | 0.76 mm | Each |
| 10600636 | 5 mL | Rheodyne Injectors 9010/9725 | 0.76 mm | Each |
| 10582075 | 50 μL | Rheodyne Injectors 7010/7125 | 0.51 mm | Each |
Product Code. 10575625
Brand: Thermo Scientific™ 8022
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Description
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Specifications
Specifications
| Type | Sample Loop |
| Description | For Rheodyne 8125; 20 uL; 0.25 mm ID |
| For Use With (Equipment) | Rheodyne Injector Model 8125 |
| Diameter (English) Inner | 0.010 in. |
| Diameter (Metric) Inner | 0.25 mm |
| Volume (Metric) | 20 μL |