Learn More
Thermo Scientific™ Rheodyne™ Sample Loops
Product Code. 10321255
Change view
Click to view available options
Volume (Metric):
2 μL
5 μL
10 μL
20 μL
50 μL
100 μL
200 μL
500 μL
1 mL
5 mL
For Use With (Equipment):
Rheodyne Injector Model 8125
Rheodyne Injectors 7010/7125
Rheodyne Injectors 7725/7725i
Rheodyne Injectors 9010/9725
Rheodyne Injectors 9725/9725i
Diameter (Metric) Inner:
0.18 mm
0.20 mm
0.25 mm
0.30 mm
0.51 mm
0.76 mm
0.8 mm, 0.5 mm, and 0.2 mm
1.0 mm
Unit Size:
Each
Product Code. Volume (Metric) For Use With (Equipment) Diameter (Metric) Inner unitSize
10321255 1 mL Rheodyne Injectors 9010/9725 0.76 mm Each
10455163 5 μL Rheodyne Injectors 7010/7125 0.18 mm Each
10621334 10 μL Rheodyne Injectors 7010/7125 0.30 mm Each
10100563 100 μL Rheodyne Injectors 7010/7125 0.51 mm Each
10776524 200 μL Rheodyne Injectors 7010/7125 0.76 mm Each
10682864 500 μL Rheodyne Injectors 7010/7125 0.76 mm Each
10465173 5 mL Rheodyne Injectors 7010/7125 1.0 mm Each
10344473 2 μL Rheodyne Injectors 9725/9725i 0.8 mm, 0.5 mm, and 0.2 mm Each
10794794 5 μL Rheodyne Injectors 7725/7725i 0.18 mm Each
10766714 10 μL Rheodyne Injectors 7725/7725i 0.30 mm Each
10466113 20 μL Rheodyne Injectors 7725/7725i 0.30 mm Each
10620184 50 μL Rheodyne Injectors 7725/7725i 0.51 mm Each
10408104 5 μL Rheodyne Injector Model 8125 0.20 mm Each
10678715 10 μL Rheodyne Injector Model 8125 0.20 mm Each
10575625 20 μL Rheodyne Injector Model 8125 0.25 mm Each
10147854 50 μL Rheodyne Injector Model 8125 0.30 mm Each
10269364 5 μL Rheodyne Injectors 9725/9725i 0.18 mm Each
10219324 10 μL Rheodyne Injectors 9725/9725i 0.25 mm Each
10555615 20 μL Rheodyne Injectors 9725/9725i 0.25 mm Each
10311255 50 μL Rheodyne Injectors 9725/9725i 0.51 mm Each
10229414 100 μL Rheodyne Injectors 9010/9725 0.51 mm Each
10651396 200 μL Rheodyne Injectors 9010/9725 0.51 mm Each
10774057 500 μL Rheodyne Injectors 9010/9725 0.76 mm Each
10600636 5 mL Rheodyne Injectors 9010/9725 0.76 mm Each
10582075 50 μL Rheodyne Injectors 7010/7125 0.51 mm Each
25 options
Product Code. 10321255
Brand: Thermo Scientific™ 9055027
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
Thermo Scientific™ Rheodyne Sample Loops are for Rheodyne sample injectors in stainless steel or biocompatible PEEK.
TRUSTED_SUSTAINABILITY
Specifications
Type Sample Loop
Description For Rheodyne 9010/9725; 1 mL; 0.76 mm ID
For Use With (Equipment) Rheodyne Injectors 9010/9725
Diameter (English) Inner 0.030 in.
Diameter (Metric) Inner 0.76 mm
Volume (Metric) 1 mL