Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Product Code. 10576005 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Rheodyne Injector 7030
Rheodyne Injector 8125
Rheodyne Injector 9010
Rheodyne Injector 9125
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7000/7010/7040
Rheodyne Injectors 7000/7010/7040/7067
Rheodyne Injectors 7060/7066
Rheodyne Injectors 7060/7066/9060
Rheodyne Injectors 7125/7126
Rheodyne Injectors 8125/8126
Material:
ETFE
PEEK
Vespel
Unit Size:
Each
Product Code. For Use With (Equipment) Material unitSize
10576005 Rheodyne Injectors 7060/7066 Vespel Each
10290285 Rheodyne Injectors 3725/3725i/3725-038/3725i-038 PEEK Each
10161263 Rheodyne Injectors 7000/7010/7040/7067 Vespel Each
10445163 Rheodyne Injectors 7000/7010/7040 ETFE Each
10671296 Rheodyne Injector 7030 Vespel Each
10648705 Rheodyne Injectors 7060/7066/9060 ETFE Each
10150893 Rheodyne Injectors 7125/7126 Vespel Each
10269514 Rheodyne Injectors 7125/7126 ETFE Each
10427534 Rheodyne Injectors 8125/8126 Vespel Each
10510126 Rheodyne Injector 8125 ETFE Each
10179464 Rheodyne Injector 9010 ETFE Each
10280845 Rheodyne Injector 9125 ETFE Each
12 options
Product Code. 10576005
Brand: Thermo Scientific™ 7060070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Maintain performance of your Thermo Scientific™ Rheodyne™ Sample Injectors for HPLC with these replacement rotor seals.
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
TRUSTED_SUSTAINABILITY
Specifications
Type Replacement Rotor Seals
Material Vespel
Description 7060-070
For Use With (Equipment) Rheodyne Injectors 7060/7066