Learn More
Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ Rheodyne™ Replacement Rotor Seals for Injectors to maintain the performance of your Rheodyne Sample Injectors for high-performance liquid chromatography (HPLC). These seals are suitable for popular Rheodyne injector models and are available in Vespel, PEEK or ETFE.
Spécification
Spécification
| Type | Replacement Rotor Seals |
| Material | Vespel |
| Description | 7125-047 |
| For Use With (Equipment) | Rheodyne Injectors 7125/7126 |