Learn More
Thermo Scientific™ RheBuild™ Kits
Product Code. 10416684 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Rheodyne Injectors 3725/3725i/3725-038/3725i-038
Rheodyne Injectors 7010/7000
Rheodyne Injectors 7125/7126
Rheodyne Injectors 7725/7725i/7726
Rheodyne Injectors 8125/8126
Rheodyne Injectors 9125/9126
Unit Size:
Each
Product Code. 10416684
Brand: Thermo Scientific™ 7125999
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Everything you need to maintain your Thermo Scientific™ Rheodyne™ valves and injectors in your HPLC system.
Use Thermo Scientific™ RheBuild Kits to maintain your Thermo Scientific™ Rheodyne™ valves and injectors for high-performance liquid chromatography (HPLC). A single kit includes everything you need.
TRUSTED_SUSTAINABILITY
Specifications
Type Maintenance Kit
Description For Rheodyne models 7125/7126
For Use With (Equipment) Rheodyne Injectors 7125/7126
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.