Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Shop All Retsch Products
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1 mm
106 μm
1.18 mm
125 μm
1.4 mm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
25 mm
Unit Size:
Each
Specifications
Specifications
| Depth (English) | 2 in. |
| Depth (Metric) | 5.08 cm |
| Pore Size | 38 μm |
| Type | Test Sieve |
| Certifications/Compliance | ASTM E11 |
| Diameter (English) Outer | 8 in. |
| Material | Stainless Steel |
| Tyler Standard | 400 Mesh |
| U.S. Standard | No. 400 |
| For Use With (Application) | Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. |
| Show More |