Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Product Code. 10278541 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1 mm
106 μm
1.18 mm
125 μm
1.4 mm
150 μm
180 μm
212 μm
250 μm
300 μm
355 μm
425 μm
500 μm
600 μm
710 μm
850 μm
25 mm
Unit Size:
Each
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10278541 425 μm Each
10014791 20 μm Each
10790463 25 μm Each
10390902 32 μm Each
10468341 38 μm Each
10534492 45 μm Each
10248971 53 μm Each
10534682 75 μm Each
10045071 106 μm Each
10085031 63 μm Each
10791423 90 μm Each
10701433 125 μm Each
10752003 212 μm Each
10585062 150 μm Each
10004921 180 μm Each
10555642 250 μm Each
10610092 300 μm Each
10024931 355 μm Each
10238591 500 μm Each
10523342 710 μm Each
10620092 600 μm Each
10208691 850 μm Each
10330622 1 mm Each
10340622 1.18 mm Each
10583722 1.4 mm Each
10136651 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Product Code. 10278541 Supplier RETSCH Supplier No. 60.150.000425
Only null left
Add to basket
Add to basket
Stainless steel with wire gauze
Specifications
Depth (English) 2 in.
Depth (Metric) 5.08 cm
Pore Size 425 μm
Type Test Sieve
Certifications/Compliance ASTM E11
Diameter (English) Outer 8 in.
Material Stainless Steel
Tyler Standard 40 Mesh
U.S. Standard No. 40
For Use With (Application) Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc.
Diameter (Metric) Outer 203 mm
Mesh Material Stainless Steel