Learn More
SI Analytics™ Resistance Thermometer with Screw Plug Head
Product Code. 10747301 Shop All SI Analytics Products
Change view
Click to view available options
Temperature Range (Metric):
-30°C to +135°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
10747301 -30°C to +135°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10747301 Supplier SI Analytics™ Supplier No. 285119162
  • ONLINE EXCLUSIVE PRICE
    3800.00 NOK
    3458.61 NOK / Pack of 1
    Save 341.39 NOK
    8% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Features a glass shaft and Pt 100 sensor
  • Screw plug head
  • Type W 2081
  • Glass shaft
  • Sensor Pt 100
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Probe 12 mm
Includes with Screw Plug Head, Type W 2081, Sensor Pt 100
Length (Metric) 120 mm
Product Type Resistance thermometer
Temperature Range (Metric) -30°C to +135°C