Description
- Use to measure temperature
- Comes equipt with plugs
- Sensors Pt 1000
Specifications
Specifications
| Diameter (Metric) Probe | 4 mm |
| Includes | 2 x 4mm banana plugs |
| Length (Metric) | 170 mm |
| Model | W 5791 NN |
| Product Type | Resistance Thermometers |
| Description | Stainless steel shaft (V4A) with sensors Pt 1000 |
| Max. Temperature (Metric) | 135°C |
| Min. Temperature (Metric) | -30°C |
| Temperature Range (Metric) | -30°C to +135°C |