missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Research™ plus BASIC - Trade-In Pipette
GREENER_CHOICE

Product Code. 15723459 Shop All Eppendorf Products
Click to view available options
Volume (Metric):
0.1 to 2.5 μL
0.25 to 2.5 mL
0.5 to 10 μL
0.5 to 5 mL
1 to 10 mL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
Color:
Blue
Dark Gray
Light Gray
Medium Gray
Orange
Red
Turquoise
Violet
Yellow
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15723459

Brand: Eppendorf™ 3123000306

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adapt for specific liquids or other external conditions with ultra-light fully autoclavable pipettes

  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejectors low operating force are positioned for ergonomics 4-digit, magnifying-shape volume display
  • Calibration display and adjustment opening for specific liquids and volumes
  • Uses renowned PhysioCare Concept™ thus reduces strain on hand and arm during pipetting, maintains balance, minimal user exertion
  • Quick lock makes lower part particularly easy to remove and service
  • Spring loaded tip cone promotes tight fit to tip
  • Has ability to be autoclaved without disassembling
  • Applications: Pipetting liquids, Mixing liquids, Filling of plates, gels, and reaction vessels, Phase extraction and removal of supernatants
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Lot Specific Certificate
Compatible Tips epT.I.P.S.
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Orange
Disposable Reusable
For Use With epT.I.P.S. 300 μL
Grip Type Ergonomic
Material Plastic
Number of Channels 1
Programmability No
Sterility Non-sterile
Type Manual Pipette
Volume (Metric) 30 to 300 μL
For Use With (Application) Pipette Tips 0.1-20 μL
For Use With (Equipment) Pipette Tips 0.5-20 μL
Includes epT.I.P.S. Pipette Tips
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.