Learn More
Rescinnamine, TRC
Product Code. 30410169
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410169 2.5 mg 2.5mg
30413642 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30410169 Supplier Toronto Research Chemicals Supplier No. TRCR1447202.5MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24815-24-5
Molecular Formula C35 H42 N2 O9
Molecular Weight (g/mol) 634.72
Synonym Yohimban-16-carboxylic acid, 11,17-dimethoxy-18-[[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]oxy]-, methyl ester, (3β,16β,17α,18β,20α)-, 3β,20α-Yohimban-16β-carboxylic acid, 18β-hydroxy-11,17α-dimethoxy-, methyl ester, 3,4,5-trimethoxycinnamate (ester) (8CI), Rescinnamine (6CI), Yohimban-16-carboxylic acid, 11,17-dimethoxy-18-[[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propenyl]oxy]-, methyl ester, (3β,16β,17α,18β,20α)- (9CI), Benz[g]indolo[2,3-a]quinolizine, yohimban-16-carboxylic acid deriv., 3,4,5-Trimethoxycinnamoyl methyl reserpate, Anaprel, Apoterin, Apoterin S, Cartric, Cinnaloid, Methyl trimethoxycinnamoylreserpate, Moderil, NSC 15628, O-(3,4,5-Trimethoxy-trans-cinnamoyl) methyl reserpate, Rescinnamin, Rescisan, Resealoid, Reserpic acid methyl ester 3,4,5-trimethoxycinnamate, Reserpinine, Reserpinine (C35 alkaloid), Scinnamina
IUPAC Name methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-[(E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoyl]oxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate
SMILES CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)\C=C\c6cc(OC)c(OC)c(OC)c6
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C35H42N2O9/c1-40-21-8-9-22-23-11-12-37-18-20-15-29(46-30(38)10-7-19-13-27(41-2)33(43-4)28(14-19)42-3)34(44-5)31(35(39)45-6)24(20)17-26(37)32(23)36-25(22)16-21/h7-10,13-14,16,20,24,26,29,31,34,36H,11-12,15,17-18H2,1-6H3/b10-7+/t20-,24+,26-,29-,31+,34+/m1/s1
Formula Weight 634.289
Chemical Name or Material Rescinnamine