Learn More
Fisherbrand™ Replacement Tips for Standard Sonic Dismembrator Probes, Models Q500 and Q700
Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
1.27 cm
1.9 cm
2.54 cm
Unit Size:
Each
Description
Requires
Cat. No. FB4207
- During normal use, tips erode and become less effective
- Easily remove and replace worn tips
- Available in 0.5, 1 and 0.75 in. diameters
- For use with Q500 and Q700 sonicators
Order Info
For replacement probes, see FB4207, FB4208. FB4209, FC4210, FB4217, FB4218, FB4219, FB4220.
Specifications
Specifications
| Product Type | Replacement Tip |
| Volume Range (Metric) | 50 to 500 mL |
| Diameter (English) | 0.75 in. |
| Diameter (English) Tip | 0.75 in. |
| Diameter (Metric) | 1.9 cm |
| Diameter (Metric) Tip | 19 mm |
| For Use With (Equipment) | 0.75 in. probe for Models Q500 and Q700 sonic dismembrators (FB4207) |
| Packaging Quantity | 1 Ea. |