Learn More
Fisherbrand™ Replacement Scoop
Scoop a variety of laboratory items with this scoop. Fisherbrand™ Traceable™ Replacement Scoops are ideal for all your scooping needs.
Brand:  Fisherbrand™
Additional Details : Weight : 0.03600kg
 View more versions of this product
Product Code. 15398764
Quantity Price
1 185.00 NOK / 40mL
10 172.09 NOK / 40mL
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
You may only order up to the available stock. For more details call customer services on +47 22 95 59 59.
Description
Description
  • A replacement scoop for your Traceable SpatulaBalance
Specifications
Specifications
Replacement Scoop
40 mL
Large
Spatula Balance