Learn More
Fisherbrand™ Replacement Scoop
Scoop a variety of laboratory items with this scoop. Fisherbrand™ Traceable™ Replacement Scoops are ideal for all your scooping needs.
185.00 NOK - 194.00 NOK
Specifications
Product Type | Replacement Scoop |
---|---|
Size | Large |
For Use With (Equipment) | Spatula Balance |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15398764
|
Fisherbrand™
|
40 mL |
185.00 NOK
40mL Qty Price Break Available |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
15388764
|
Fisherbrand™
|
30 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- A replacement scoop for your Traceable SpatulaBalance
Specifications
Replacement Scoop | |
Large | |
Spatula Balance |