Learn More
Eppendorf™ Replacement Rotor Lids
Shop All Eppendorf Products
Click to view available options
For Use With (Equipment):
F-35-6-30 Rotor
FA-45-12-17 Rotor
Unit Size:
Each
Description
- Replacement Rotor Lids
- Available for a wide range of products
Specifications
Specifications
| Type | Non Aerosol-tight |
| Material | Polypropylene |
| For Use With (Equipment) | F-35-6-30 Rotor |
| Color | Black |
| Description | Rotor lid for F-35-6-30 |