Learn More
Fisherbrand™ Replacement Pen Cartridge
Product Code. 13128325 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Black
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
13128325 Black Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13128325
  • ONLINE EXCLUSIVE PRICE
    107.00 NOK
    88.09 NOK / Pack of 1
    Save 18.91 NOK
    17% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
Sound and count occur simultaneously when inserted correctly into Counter-Pen™. Fisherbrand™ Replacement Pen Cartridge marks with black felt tip.
Specifications
Tip Style Felt Tip
Product Type Replacement Cartridge for Counter-pen
Color Black
For Use With (Equipment) Counter-Pen™