Learn More
Fisherbrand™ Replacement Part Kit for Heat Sealer
Product Code. 15350407 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15350407 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15350407
  • ONLINE EXCLUSIVE PRICE
    337.00 NOK
    247.93 NOK / Pack of 1
    Save 89.07 NOK
    26% Off
View all promotions
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
Replace burnt out heating elements and dull blades. Replacement Part Kit for Heat Sealer also includes 2 PTFE cloths.
  • For use with Fisherbrand™ Heat Sealer with Cutter.
  • Kit includes:
    • 2 blades
    • 2 heating elements
    • 2 PTFE cloths
Specifications
Product Type Heat Sealer
For Use With (Equipment) Heat Sealer and Cutter model HS28270
Includes 2 replacement blades, 2 heating elements, and 2 PTFE cloths
Quantity 1 Ea.