Learn More
Gilson™ REPETMAN™ Non-Sterile Tips
Shop All Gilson Products
Click to view available options
Volume (Metric):
1.0 mL
10 mL
Unit Size:
Pack of 100
Description
- Non-sterile
- Available in 1 and 10mL volumes
- Aids in positive displacement pipetting
- Provided in packs of 100
Specifications
Specifications
| Description | Pipette tips |
| Volume (Metric) | 10 mL |
| Sterility | Non-sterile |
| For Use With (Application) | Pipetting |
| Tip Style | Positive Displacement |
| Quantity | 100/pk |
| For Use With (Equipment) | Repetman |
| Product Line | Repetman |