Description
- With guide bars and fixing screws
- Requires option K1
- Consider max. loading of chamber; max. loading 60 kg
Specifications
Specifications
| For Use With (Equipment) | IPP750eco |
| Material | Stainless Steel |