Learn More
Hanna Instruments™ Refillable Digital pH Electrode
Product Code. 15178239
Change view
Click to view available options
Includes:
No Matching Pin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
15178239 No Matching Pin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15178239 Supplier Hanna Instruments™ Supplier No. HI11310
  • ONLINE EXCLUSIVE PRICE
    5663.36 NOK
    3710.75 NOK / Pack of 1
    Save 1952.61 NOK
    34% Off
View all promotions
Estimated Shipment: 17-11-2026
Specifications
Connector Types 3.5 mm Jack
Junction Ceramic Single, 15 to 20 μL/h
Material Glass
pH Range 0 to 13
Temperature (Metric) Operating -5°C to +100°C
Type pH Electrode
Includes No Matching Pin
Length (Metric) Cable 1 m
Diameter (Metric) Cap 9.5 mm