Learn More
Recoverin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10073-1-AP-150UL
 View more versions of this product
Product Code. 16846223
  • 4836.10 NOK / 150µL
Description
Calcium binding proteins are implicated as being a helper molecule in aberrant tissue growth of tumors. Recoverin is a neurologically important calcium binding protein, involved in tumor pathology. It is conserved across species and there are indications of several isoforms of this molecule. Recoverin antibodies are a useful tool for distinguishing between pineal gland-derived tissue and the surrounding tissue of the brain. They can also be used to accurately determine maligt tissue in this location of the brain. In pineal tumor tissue, the normal tissue staining pattern is lost, indicating the tissue′s loss of this protein.
Specifications
Specifications
Recoverin
Polyclonal
Unconjugated
RCVRN
Protein CAR, RCV1, RCVRN, recoverin
Rabbit
Antigen Affinity Chromatography
RUO
140936, 19674, 5957
-20°C
Liquid
Immunofluorescence, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P34057, P35243
RCVRN
Recoverin Fusion Protein Ag0119
150 μL
Primary
Human, Mouse, Rat