missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Sheep IL12B / P40 Protein

A DNA sequence encoding the Ovis aries (Sheep) IL12B / P40, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6210-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011600

  • 41115.00 NOK / 1mg
Estimated Shipment: 11-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001009438.1
IL12B / P40 Protein
Research Use Only
IL12B
Recombinant Protein
Yeast
IWELEKNVYV VELDWYPNAP GETVVLTCDT PEEDGITWTS DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSRSLL LLHKKEDGIW STDILKDQKE PKAKSFLKCE AKDYSGHFTC SWLTAISTNL KFSVKSSRGS SDPRGVTCGA ASLSAEKVSM DHREYNKYTV ECQEGSACPA AEESLPIEVV MEAVHKLKYE NYTSSFFIRD IIKPDPPKNL QLRPLKNSRQ VEVSWEYPDT WSTPHSYFSL TFCVQVQGKN KREKKLFTDQ TSAKVTCHKD ANIRVQARDR YYSSFWSEWA SVSCS
Greater than 90% as determined by SDS-PAGE.
443472
1 mg
Not determined.
Full Length
Sheep
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers