missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL12B / P40 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL12B / P40, was expressed in the hosts and tags indicated.
3633.50 NOK - 41115.00 NOK
Specifications
Accession Number | NP_001009438.1 |
---|---|
Gene ID (Entrez) | 443472 |
Name | IL12B / P40 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091590
|
enQuireBio™
QP6210-YE-100UG |
100 μg |
11060.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011600
|
enQuireBio™
QP6210-YE-1MG |
1 mg |
41115.00 NOK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001600
|
enQuireBio™
QP6210-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021600
|
enQuireBio™
QP6210-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031600
|
enQuireBio™
QP6210-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041600
|
enQuireBio™
QP6210-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001009438.1 | |
IL12B / P40 Protein | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |
443472 | |
Research Use Only | |
IL12B | |
Recombinant Protein | |
Yeast | |
IWELEKNVYV VELDWYPNAP GETVVLTCDT PEEDGITWTS DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSRSLL LLHKKEDGIW STDILKDQKE PKAKSFLKCE AKDYSGHFTC SWLTAISTNL KFSVKSSRGS SDPRGVTCGA ASLSAEKVSM DHREYNKYTV ECQEGSACPA AEESLPIEVV MEAVHKLKYE NYTSSFFIRD IIKPDPPKNL QLRPLKNSRQ VEVSWEYPDT WSTPHSYFSL TFCVQVQGKN KREKKLFTDQ TSAKVTCHKD ANIRVQARDR YYSSFWSEWA SVSCS | |
Greater than 90% as determined by SDS-PAGE. |