missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Osteopontin / SPP1 / ETA-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Osteopontin / SPP1 / ETA-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6722-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001191162.1 | |
Osteopontin / SPP1 / ETA-1 Protein | |
Research Use Only | |
Spp1 | |
Recombinant Protein | |
Yeast | |
LPVKVTDSGS SEEKLYSLHP DPIATWLVPD PSQKQNLLAP QNAVSSEEKD DFKQETLPSN SNESHDHMDD DDDDDDDDGD HAESEDSVDS DESDESHHSD ESDETVTAST QADTFTPIVP TVDVPNGRGD SLAYGLRSKS RSFQVSDEQY PDATDEDLTS HMKSGESKES LDVIPVAQLL SMPSDQDNNG KGSHESSQLD EPSLETHRLE HSKESQESAD QSDVIDSQAS SKASLEHQSH KFHSHKDKLV LDPKSKEDDR YLKFRISHEL ESSS | |
Greater than 90% as determined by SDS-PAGE. |
20750 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |