missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Osteopontin / SPP1 / ETA-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Osteopontin / SPP1 / ETA-1, was expressed in the hosts and tags indicated.
2526.25 NOK - 27321.13 NOK
Specifications
Accession Number | NP_001191162.1 |
---|---|
Gene ID (Entrez) | 20750 |
Name | Osteopontin / SPP1 / ETA-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085380
|
enQuireBio™
QP6722-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025390
|
enQuireBio™
QP6722-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075380
|
enQuireBio™
QP6722-YE-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005390
|
enQuireBio™
QP6722-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015390
|
enQuireBio™
QP6722-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095380
|
enQuireBio™
QP6722-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001191162.1 | |
Osteopontin / SPP1 / ETA-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20750 | |
Research Use Only | |
Spp1 | |
Recombinant Protein | |
Yeast | |
LPVKVTDSGS SEEKLYSLHP DPIATWLVPD PSQKQNLLAP QNAVSSEEKD DFKQETLPSN SNESHDHMDD DDDDDDDDGD HAESEDSVDS DESDESHHSD ESDETVTAST QADTFTPIVP TVDVPNGRGD SLAYGLRSKS RSFQVSDEQY PDATDEDLTS HMKSGESKES LDVIPVAQLL SMPSDQDNNG KGSHESSQLD EPSLETHRLE HSKESQESAD QSDVIDSQAS SKASLEHQSH KFHSHKDKLV LDPKSKEDDR YLKFRISHEL ESSS | |
Greater than 90% as determined by SDS-PAGE. |