missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein
A DNA sequence encoding the Mus musculus (Mouse) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.
2526.25 NOK - 27321.13 NOK
Specifications
Accession Number | NP_001295215.1 |
---|---|
Gene ID (Entrez) | 14198 |
Name | Fragile histidine triad / FHIT Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069040
|
enQuireBio™
QP7503-YE-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16009050
|
enQuireBio™
QP7503-YE-500UG |
500 μg |
22140.00 NOK
500µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079040
|
enQuireBio™
QP7503-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099040
|
enQuireBio™
QP7503-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019050
|
enQuireBio™
QP7503-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089040
|
enQuireBio™
QP7503-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001295215.1 | |
Fragile histidine triad / FHIT Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
14198 | |
Research Use Only | |
Fhit | |
Recombinant Protein | |
Yeast | |
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFRDLH PDEVADLFQV TQRVGTVVEK HFQGTSITFS MQDGPEAGQT VKHVHVHVLP RKAGDFPRND NIYDELQKHD REEEDSPAFW RSEKEMAAEA EALRVYFQA | |
Greater than 90% as determined by SDS-PAGE. |