missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fragile histidine triad / FHIT Protein
A DNA sequence encoding the Mus musculus (Mouse) Fragile histidine triad / FHIT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7503-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001295215.1 | |
Fragile histidine triad / FHIT Protein | |
Research Use Only | |
Fhit | |
Recombinant Protein | |
Yeast | |
SFRFGQHLIK PSVVFLKTEL SFALVNRKPV VPGHVLVCPL RPVERFRDLH PDEVADLFQV TQRVGTVVEK HFQGTSITFS MQDGPEAGQT VKHVHVHVLP RKAGDFPRND NIYDELQKHD REEEDSPAFW RSEKEMAAEA EALRVYFQA | |
Greater than 90% as determined by SDS-PAGE. |
14198 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |