Learn More
enQuireBio™ Recombinant Mouse FGF21 / Fibroblast Growth Factor 21 Protein
A DNA sequence encoding the Mus musculus (Mouse) FGF21 / Fibroblast Growth Factor 21, was expressed in the hosts and tags indicated.
3160.00 NOK - 27321.13 NOK
Specifications
Accession Number | NP_064397.1 |
---|---|
Gene ID (Entrez) | 56636 |
Name | FGF21 / Fibroblast Growth Factor 21 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15909569
|
enQuireBio™
QP6044-EC-10UG |
10 μg |
3160.00 NOK
10µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999559
|
enQuireBio™
QP6044-EC-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939569
|
enQuireBio™
QP6044-EC-500UG |
500 μg |
22265.00 NOK
500µg |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15929569
|
enQuireBio™
QP6044-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15949569
|
enQuireBio™
QP6044-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919569
|
enQuireBio™
QP6044-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_064397.1 | |
FGF21 / Fibroblast Growth Factor 21 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56636 | |
Research Use Only | |
FGF21 | |
Recombinant Protein | |
E. coli | |
AYPIPDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGAAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GALYGSPHFD PEACSFRELL LEDGYNVYQS EAHGLPLRLP QKDSPNQDAT SWGPVRFLPM PGLLHEPQDQ AGFLPPEPPD VGSSDPLSMV EPLQGRSPSY AS | |
Greater than 90% as determined by SDS-PAGE. |