missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL3 / GRO gamma Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.
2526.25 NOK - 34365.00 NOK
Specifications
Accession Number | NP_976065.1 |
---|---|
Gene ID (Entrez) | 330122 |
Name | CXCL3 / GRO gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16085771
|
enQuireBio™
QP8652-EC-100UG |
100 μg |
7745.00 NOK
100µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005781
|
enQuireBio™
QP8652-EC-1MG |
1 mg |
34365.00 NOK
1mg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16095771
|
enQuireBio™
QP8652-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015781
|
enQuireBio™
QP8652-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16025781
|
enQuireBio™
QP8652-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16035781
|
enQuireBio™
QP8652-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_976065.1 | |
CXCL3 / GRO gamma Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
330122 | |
Research Use Only | |
CXCL3 | |
Recombinant Protein | |
E. coli | |
SELRCQCLNT LPRVDFETIQ SLTVTPPGPH CTQTEVIATL KDGQEVCLNP QGPRLQIIIK KILKSGKSS | |
Greater than 90% as determined by SDS-PAGE. |