missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL3 / GRO gamma Protein
A DNA sequence encoding the Mus musculus (Mouse) CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8652-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_976065.1 | |
CXCL3 / GRO gamma Protein | |
Research Use Only | |
CXCL3 | |
Recombinant Protein | |
E. coli | |
SELRCQCLNT LPRVDFETIQ SLTVTPPGPH CTQTEVIATL KDGQEVCLNP QGPRLQIIIK KILKSGKSS | |
Greater than 90% as determined by SDS-PAGE. |
330122 | |
200 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |