missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPF0454 protein C12orf49 Protein
A DNA sequence encoding the Homo sapiens (Human) UPF0454 protein C12orf49, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8088-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_079014.1 | |
UPF0454 protein C12orf49 Protein | |
Research Use Only | |
C12orf49 | |
Recombinant Protein | |
E. coli | |
TFKQEERAVR DRNLLQVHDH NQPIPWKVQF NLGNSSRPSN QCRNSIQGKH LITDELGYVC ERKDLLVNGC CNVNVPSTKQ YCCDGCWPNG CCSAYEYCVS CCLQPNKQLL LERFLNRAAV AFQNLFMAVE DHFELCLAKC RTSSQSVQHE NTYRDPIAKY CYGESPPELF PA | |
Greater than 90% as determined by SDS-PAGE. |
79794 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |