missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UPF0454 protein C12orf49 Protein
A DNA sequence encoding the Homo sapiens (Human) UPF0454 protein C12orf49, was expressed in the hosts and tags indicated.
1644.75 NOK - 24050.00 NOK
Specifications
Accession Number | NP_079014.1 |
---|---|
Gene ID (Entrez) | 79794 |
Name | UPF0454 protein C12orf49 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002601
|
enQuireBio™
QP8088-EC-100UG |
100 μg |
5325.00 NOK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16022601
|
enQuireBio™
QP8088-EC-1MG |
1 mg |
24050.00 NOK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16012601
|
enQuireBio™
QP8088-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16032601
|
enQuireBio™
QP8088-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16042601
|
enQuireBio™
QP8088-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052601
|
enQuireBio™
QP8088-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_079014.1 | |
UPF0454 protein C12orf49 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79794 | |
Research Use Only | |
C12orf49 | |
Recombinant Protein | |
E. coli | |
TFKQEERAVR DRNLLQVHDH NQPIPWKVQF NLGNSSRPSN QCRNSIQGKH LITDELGYVC ERKDLLVNGC CNVNVPSTKQ YCCDGCWPNG CCSAYEYCVS CCLQPNKQLL LERFLNRAAV AFQNLFMAVE DHFELCLAKC RTSSQSVQHE NTYRDPIAKY CYGESPPELF PA | |
Greater than 90% as determined by SDS-PAGE. |