missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine--tRNA ligase, cytoplasmic Protein
A DNA sequence encoding the Homo sapiens (Human) Serine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8468-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006504.2 | |
Serine--tRNA ligase, cytoplasmic Protein | |
Research Use Only | |
SERS | |
Recombinant Protein | |
E. coli | |
VLDLDLFRVD KGGDPALIRE TQEKRFKDPG LVDQLVKADS EWRRCRFRAD NLNKLKNLCS KTIGEKMKKK EPVGDDESVP ENVLSFDDLT ADALANLKVS QIKKVRLLID EAILKCDAER IKLEAERFEN LREIGNLLHP SVPISNDEDV DNKVERIWGD CTVRKKYSHV DLVVMVDGFE GEKGAVVAGS RGYFLKGVLV FLEQALIQYA LRTLGSRGYI PIYTPFFMRK EV | |
Greater than 90% as determined by SDS-PAGE. |
6301 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |