missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine--tRNA ligase, cytoplasmic Protein
A DNA sequence encoding the Homo sapiens (Human) Serine--tRNA ligase, cytoplasmic, was expressed in the hosts and tags indicated.
1644.75 NOK - 19129.63 NOK
Specifications
Accession Number | NP_006504.2 |
---|---|
Gene ID (Entrez) | 6301 |
Name | Serine--tRNA ligase, cytoplasmic Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094771
|
enQuireBio™
QP8468-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034781
|
enQuireBio™
QP8468-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084771
|
enQuireBio™
QP8468-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014781
|
enQuireBio™
QP8468-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024781
|
enQuireBio™
QP8468-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004781
|
enQuireBio™
QP8468-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006504.2 | |
Serine--tRNA ligase, cytoplasmic Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6301 | |
Research Use Only | |
SERS | |
Recombinant Protein | |
E. coli | |
VLDLDLFRVD KGGDPALIRE TQEKRFKDPG LVDQLVKADS EWRRCRFRAD NLNKLKNLCS KTIGEKMKKK EPVGDDESVP ENVLSFDDLT ADALANLKVS QIKKVRLLID EAILKCDAER IKLEAERFEN LREIGNLLHP SVPISNDEDV DNKVERIWGD CTVRKKYSHV DLVVMVDGFE GEKGAVVAGS RGYFLKGVLV FLEQALIQYA LRTLGSRGYI PIYTPFFMRK EV | |
Greater than 90% as determined by SDS-PAGE. |