missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS27A Protein
A cDNA sequence encoding the RPS27A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13356-B-EC-10ug
Additional Details : Weight : 0.01000kg
Specifications
7311; 7314; 6233; 7316 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
The Ubiquitin protein solution contains 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
RPS27A Protein | |
Research Use Only | |
RPS27A; UBB; UBC; UBA52 | |
Human | |
Biotin |