missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RPS27A Protein
A cDNA sequence encoding the RPS27A was constructed and used to recombinantly synthesize the protein.
2140.00 NOK - 27485.00 NOK
Specifications
Gene ID (Entrez) | 7311; 7314; 6233; 7316 |
---|---|
Name | RPS27A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | RPS27A; UBB; UBC; UBA52 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973309
|
enQuireBio™
QP13356-B-EC-10UG |
10 μg |
2140.00 NOK
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953309
|
enQuireBio™
QP13356-50UG |
50 μg |
2140.00 NOK
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15943309
|
enQuireBio™
QP13356-1MG |
1 mg |
27485.00 NOK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7311; 7314; 6233; 7316 | |
Research Use Only | |
RPS27A; UBB; UBC; UBA52 | |
Human | |
The Ubiquitin protein solution contains 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |
RPS27A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli |