missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nephrin Protein
A DNA sequence encoding the Homo sapiens (Human) Nephrin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6429-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004637.1 | |
Nephrin Protein | |
Research Use Only | |
NPHS1 | |
Recombinant Protein | |
E. coli | |
QLAIPASVPR GFWALPENLT VVEGASVELR CGVSTPGSAV QWAKDGLLLG PDPRIPGFPR YRLEGDPARG EFHLHIEACD LSDDAEYECQ VGRSEMGPEL VSPRVILSIL VPPKLLLLTP EAGTMVTWVA GQEYVVNCVS GDAKPAPDIT ILLSGQTISD ISANVNEGSQ QKLFTVEATA RVTPRSSDNR QLLVCEASSP ALEAPIKASF TVNVLFPPGP PVIEWPGLDE GHVRA | |
Greater than 90% as determined by SDS-PAGE. |
4868 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |