missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Nephrin Protein
A DNA sequence encoding the Homo sapiens (Human) Nephrin, was expressed in the hosts and tags indicated.
2040.00 NOK - 23690.00 NOK
Specifications
Accession Number | NP_004637.1 |
---|---|
Gene ID (Entrez) | 4868 |
Name | Nephrin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003250
|
enQuireBio™
QP6429-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043250
|
enQuireBio™
QP6429-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093240
|
enQuireBio™
QP6429-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023250
|
enQuireBio™
QP6429-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033250
|
enQuireBio™
QP6429-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013250
|
enQuireBio™
QP6429-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004637.1 | |
Nephrin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4868 | |
Research Use Only | |
NPHS1 | |
Recombinant Protein | |
E. coli | |
QLAIPASVPR GFWALPENLT VVEGASVELR CGVSTPGSAV QWAKDGLLLG PDPRIPGFPR YRLEGDPARG EFHLHIEACD LSDDAEYECQ VGRSEMGPEL VSPRVILSIL VPPKLLLLTP EAGTMVTWVA GQEYVVNCVS GDAKPAPDIT ILLSGQTISD ISANVNEGSQ QKLFTVEATA RVTPRSSDNR QLLVCEASSP ALEAPIKASF TVNVLFPPGP PVIEWPGLDE GHVRA | |
Greater than 90% as determined by SDS-PAGE. |