missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin-26 Protein
A DNA sequence encoding the Homo sapiens (Human) Interleukin-26, was expressed in the hosts and tags indicated.
2085.50 NOK - 22225.63 NOK
Specifications
Accession Number | NP_060872.1 |
---|---|
Gene ID (Entrez) | 55801 |
Name | Interleukin-26 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068961
|
enQuireBio™
QP9904-YE-100UG |
100 μg |
6475.00 NOK
100µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008971
|
enQuireBio™
QP9904-YE-500UG |
500 μg |
17555.00 NOK
500µg |
Estimated Shipment: 01-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078961
|
enQuireBio™
QP9904-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098961
|
enQuireBio™
QP9904-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018971
|
enQuireBio™
QP9904-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088961
|
enQuireBio™
QP9904-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_060872.1 | |
Interleukin-26 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55801 | |
Research Use Only | |
IL26 | |
Recombinant Protein | |
Yeast | |
KHKQSSFTKS CYPRGTLSQA VDALYIKAAW LKATIPEDRI KNIRLLKKKT KKQFMKNCQF QEQLLSFFME DVFGQLQLQG CKKIRFVEDF HSLRQKLSHC ISCASSAREM KSITRMKRIF YRIGNKGIYK AISELDILLS WIKKLLESSQ | |
Greater than 90% as determined by SDS-PAGE. |