missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interleukin-26 Protein
A DNA sequence encoding the Homo sapiens (Human) Interleukin-26, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9904-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_060872.1 | |
Interleukin-26 Protein | |
Research Use Only | |
IL26 | |
Recombinant Protein | |
Yeast | |
KHKQSSFTKS CYPRGTLSQA VDALYIKAAW LKATIPEDRI KNIRLLKKKT KKQFMKNCQF QEQLLSFFME DVFGQLQLQG CKKIRFVEDF HSLRQKLSHC ISCASSAREM KSITRMKRIF YRIGNKGIYK AISELDILLS WIKKLLESSQ | |
Greater than 90% as determined by SDS-PAGE. |
55801 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |