missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2B type 1-B Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2B type 1-B, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6159-ec-500ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_066406.1 | |
Histone H2B type 1-B Protein | |
Research Use Only | |
HIST1H2BB | |
Recombinant Protein | |
E. coli | |
PEPSKSAPAP KKGSKKAITK AQKKDGKKRK RSRKESYSIY VYKVLKQVHP DTGISSKAMG IMNSFVNDIF ERIAGEASRL AHYNKRSTIT SREIQTAVRL LLPGELAKHA VSEGTKAVTK YTSSK | |
Greater than 90% as determined by SDS-PAGE. |
3018 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |